siRNA Details
| Gene Symbol: Gnptab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004164.2 | NP_001004164.2 | 15 | 3066 | ||
| GenBank mRNA | AK140867.1 | - | 14 | 3170 | ||
| AK154710.1 | - | 7 | 1965 | |||
| AK173132.1 | - | 15 | 3066 | |||
| BC060638.1 | - | 17 | 2550 | 6849405 | ||

Gnptab
Genomic Map