siRNA Details
| Gene Symbol: Gnptab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004164.2 | NP_001004164.2 | 15, 16 | 3087 | ||
| GenBank mRNA | AK140867.1 | - | 14, 15 | 3191 | ||
| AK154710.1 | - | 7, 8 | 1986 | |||
| AK173132.1 | - | 15, 16 | 3087 | |||
| BC060638.1 | - | 17, 18 | 2571 | 6849405 | ||
| L36434.1 | - | 3, 4 | 1156 | |||

Gnptab
Genomic Map