siRNA Details
| Gene Symbol: Sgpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004173.2 | NP_001004173.1 | 5 | 1011 | ||
| GenBank mRNA | AK081023.1 | - | 1 | 867 | ||
| BC049995.1 | - | 5 | 1012 | 6399201 | ||
| BC147843.1 | - | 5 | 1200 | 9087560 | ||
| BC147850.1 | - | 5 | 1200 | 9087567 | ||

Sgpp2
Genomic Map