siRNA Details
| Gene Symbol: Sgpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004173.2 | NP_001004173.1 | 5 | 1698 | ||
| GenBank mRNA | AK081023.1 | - | 1 | 1554 | ||
| BC049995.1 | - | 5 | 1699 | 6399201 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgpp2| Gene Symbol: Sgpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004173.2 | NP_001004173.1 | 5 | 1698 | ||
| GenBank mRNA | AK081023.1 | - | 1 | 1554 | ||
| BC049995.1 | - | 5 | 1699 | 6399201 | ||