siRNA Details
| Gene Symbol: Fam222a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004180.1 | NP_001004180.1 | 3 | 1405 | ||
| GenBank mRNA | BC057022.1 | - | 3 | 1405 | 6813454 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam222a| Gene Symbol: Fam222a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004180.1 | NP_001004180.1 | 3 | 1405 | ||
| GenBank mRNA | BC057022.1 | - | 3 | 1405 | 6813454 | |