siRNA Details
Gene Symbol: KIAA0125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026800.2 | 6 | 8787 | |||
GenBank mRNA | BC152473.1 | - | 1 | 7060 | ||
D50915.1 | - | 1 | 7060 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KIAA0125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026800.2 | 6 | 8787 | |||
GenBank mRNA | BC152473.1 | - | 1 | 7060 | ||
D50915.1 | - | 1 | 7060 |