siRNA Details
| Gene Symbol: Zfp560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004190.3 | NP_001004190.2 | 7 | 2533 | ||
| GenBank mRNA | AK028777.1 | - | 1 | 1112 | ||
| AK141494.1 | - | 7 | 2533 | |||
| BC141211.1 | - | 7 | 2520 | 9055823 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp560| Gene Symbol: Zfp560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004190.3 | NP_001004190.2 | 7 | 2533 | ||
| GenBank mRNA | AK028777.1 | - | 1 | 1112 | ||
| AK141494.1 | - | 7 | 2533 | |||
| BC141211.1 | - | 7 | 2520 | 9055823 | ||