siRNA Details
| Gene Symbol: Tax1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004199.1 | NP_001004199.1 | 4, 5 | 545 | ||
| XM_006236480.2 | XP_006236542.1 | 4, 5 | 588 | |||
| XM_006236481.2 | XP_006236543.1 | 4, 5 | 1031 | |||
| XM_006236482.2 | XP_006236544.1 | 4, 5 | 588 | |||
| XM_006236483.2 | XP_006236545.1 | 4, 5 | 588 | |||
| GenBank mRNA | AY318959.1 | - | 461 | |||
| BC081949.1 | - | 4, 5 | 545 | 7122283 | ||

Tax1bp1
Genomic Map