siRNA Details
| Gene Symbol: Ccl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC079460.1 | - | 1221 | 7121977 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl6| Gene Symbol: Ccl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC079460.1 | - | 1221 | 7121977 | ||