siRNA Details
| Gene Symbol: Rilpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004205.1 | NP_001004205.1 | 4 | 944 | ||
| GenBank mRNA | BC079132.1 | - | 4 | 944 | 7127390 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rilpl2| Gene Symbol: Rilpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004205.1 | NP_001004205.1 | 4 | 944 | ||
| GenBank mRNA | BC079132.1 | - | 4 | 944 | 7127390 | |