siRNA Details
| Gene Symbol: Dhps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004207.1 | NP_001004207.1 | 8 | 980 | ||
| GenBank mRNA | BC079188.1 | - | 8 | 980 | 7131199 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhps| Gene Symbol: Dhps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004207.1 | NP_001004207.1 | 8 | 980 | ||
| GenBank mRNA | BC079188.1 | - | 8 | 980 | 7131199 | |