siRNA Details
Gene Symbol: Cacybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004208.1 | NP_001004208.1 | 6 | 1310 | ||
GenBank mRNA | BC079007.1 | - | 6 | 1310 | 7113129 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cacybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004208.1 | NP_001004208.1 | 6 | 1310 | ||
GenBank mRNA | BC079007.1 | - | 6 | 1310 | 7113129 |