siRNA Details
| Gene Symbol: Saraf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004213.1 | NP_001004213.1 | 6 | 1678 | ||
| GenBank mRNA | BC078979.1 | - | 6 | 1678 | 7112025 | |
| FQ211074.1 | - | 1 | 521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Saraf| Gene Symbol: Saraf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004213.1 | NP_001004213.1 | 6 | 1678 | ||
| GenBank mRNA | BC078979.1 | - | 6 | 1678 | 7112025 | |
| FQ211074.1 | - | 1 | 521 | |||