siRNA Details
| Gene Symbol: Emc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004221.1 | NP_001004221.1 | 7 | 1492 | ||
| XM_006229000.2 | XP_006229062.1 | 8 | 1595 | |||
| GenBank mRNA | BC079041.1 | - | 7 | 1492 | 7114835 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emc10| Gene Symbol: Emc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004221.1 | NP_001004221.1 | 7 | 1492 | ||
| XM_006229000.2 | XP_006229062.1 | 8 | 1595 | |||
| GenBank mRNA | BC079041.1 | - | 7 | 1492 | 7114835 | |