siRNA Details
| Gene Symbol: Rtcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004227.1 | NP_001004227.1 | 11 | 1117 | ||
| XM_008761456.1 | XP_008759678.1 | 10 | 1073 | |||
| GenBank mRNA | BC079374.1 | - | 11 | 1117 | 7110129 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtcd1| Gene Symbol: Rtcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004227.1 | NP_001004227.1 | 11 | 1117 | ||
| XM_008761456.1 | XP_008759678.1 | 10 | 1073 | |||
| GenBank mRNA | BC079374.1 | - | 11 | 1117 | 7110129 | |