siRNA Details
| Gene Symbol: Emcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004228.1 | NP_001004228.1 | 3 | 269 | ||
| XM_006233366.2 | XP_006233428.1 | 3 | 354 | |||
| GenBank mRNA | BC079155.1 | - | 3 | 269 | 7128709 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emcn| Gene Symbol: Emcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004228.1 | NP_001004228.1 | 3 | 269 | ||
| XM_006233366.2 | XP_006233428.1 | 3 | 354 | |||
| GenBank mRNA | BC079155.1 | - | 3 | 269 | 7128709 | |