siRNA Details
| Gene Symbol: Edem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004230.1 | NP_001004230.1 | 11 | 1340 | ||
| GenBank mRNA | BC079029.1 | - | 11 | 1340 | 7114041 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Edem2| Gene Symbol: Edem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004230.1 | NP_001004230.1 | 11 | 1340 | ||
| GenBank mRNA | BC079029.1 | - | 11 | 1340 | 7114041 | |