siRNA Details
| Gene Symbol: Fscn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004232.1 | NP_001004232.1 | 2 | 426 | ||
| GenBank mRNA | BC079410.1 | - | 2 | 426 | 7113273 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fscn3| Gene Symbol: Fscn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004232.1 | NP_001004232.1 | 2 | 426 | ||
| GenBank mRNA | BC079410.1 | - | 2 | 426 | 7113273 | |