siRNA Details
Gene Symbol: Fscn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004232.1 | NP_001004232.1 | 3 | 976 | ||
GenBank mRNA | BC079410.1 | - | 3 | 976 | 7113273 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fscn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004232.1 | NP_001004232.1 | 3 | 976 | ||
GenBank mRNA | BC079410.1 | - | 3 | 976 | 7113273 |