siRNA Details
| Gene Symbol: Mkrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004233.1 | NP_001004233.1 | 5 | 1078 | ||
| GenBank mRNA | BC079407.1 | - | 5 | 1078 | 7112867 | |
| FQ212797.1 | - | 3 | 883 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mkrn1| Gene Symbol: Mkrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004233.1 | NP_001004233.1 | 5 | 1078 | ||
| GenBank mRNA | BC079407.1 | - | 5 | 1078 | 7112867 | |
| FQ212797.1 | - | 3 | 883 | |||