siRNA Details
| Gene Symbol: Uqcrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004250.2 | NP_001004250.1 | 13 | 1544 | ||
| GenBank mRNA | BC078923.1 | - | 13 | 1546 | 7106265 | |
| FQ210217.1 | - | 6 | 663 | |||
| FQ213124.1 | - | 13 | 1555 | |||
| FQ215592.1 | - | 9 | 1028 | |||
| FQ216746.1 | - | 13 | 1548 | |||
| FQ224157.1 | - | 13 | 1552 | |||
| FQ229052.1 | - | 13 | 1571 | |||

Uqcrc1
Genomic Map