siRNA Details
| Gene Symbol: Uqcrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004250.2 | NP_001004250.1 | 4 | 391 | ||
| GenBank mRNA | BC078923.1 | - | 4 | 393 | 7106265 | |
| FQ213124.1 | - | 4 | 398 | |||
| FQ216746.1 | - | 4 | 394 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uqcrc1| Gene Symbol: Uqcrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004250.2 | NP_001004250.1 | 4 | 391 | ||
| GenBank mRNA | BC078923.1 | - | 4 | 393 | 7106265 | |
| FQ213124.1 | - | 4 | 398 | |||
| FQ216746.1 | - | 4 | 394 | |||