siRNA Details
Gene Symbol: MGC94335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004251.1 | NP_001004251.1 | 5 | 1351 | ||
GenBank mRNA | BC079246.1 | - | 5 | 1351 | 7132862 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MGC94335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004251.1 | NP_001004251.1 | 5 | 1351 | ||
GenBank mRNA | BC079246.1 | - | 5 | 1351 | 7132862 |