siRNA Details
| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004252.1 | NP_001004252.1 | 5 | 387 | ||
| XM_006245189.2 | XP_006245251.1 | 5 | 423 | |||
| GenBank mRNA | BC079364.1 | - | 5 | 387 | 7109296 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Farsb| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004252.1 | NP_001004252.1 | 5 | 387 | ||
| XM_006245189.2 | XP_006245251.1 | 5 | 423 | |||
| GenBank mRNA | BC079364.1 | - | 5 | 387 | 7109296 | |