siRNA Details
| Gene Symbol: Sirt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004256.1 | NP_001004256.1 | 8 | 1181 | ||
| XM_006253801.1 | XP_006253863.1 | 9 | 1365 | |||
| XM_006253802.1 | XP_006253864.1 | 9 | 1243 | |||
| XM_006253803.2 | XP_006253865.1 | 9 | 1237 | |||
| XM_006253804.2 | XP_006253866.1 | 9 | 1188 | |||
| GenBank mRNA | BC078958.1 | - | 8 | 1181 | 7110841 | |

Sirt5
Genomic Map