siRNA Details
Gene Symbol: Cog6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004262.1 | NP_001004262.1 | 4, 5 | 424 | ||
GenBank mRNA | BC079442.1 | - | 4, 5 | 424 | 7115484 | |
FQ225755.1 | - | 215 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cog6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004262.1 | NP_001004262.1 | 4, 5 | 424 | ||
GenBank mRNA | BC079442.1 | - | 4, 5 | 424 | 7115484 | |
FQ225755.1 | - | 215 |