siRNA Details
Gene Symbol: Tmem106b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004267.1 | NP_001004267.1 | 6 | 670 | ||
GenBank mRNA | AY387087.1 | - | 873 | |||
BC079127.1 | - | 6 | 670 | 7127112 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem106b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004267.1 | NP_001004267.1 | 6 | 670 | ||
GenBank mRNA | AY387087.1 | - | 873 | |||
BC079127.1 | - | 6 | 670 | 7127112 |