siRNA Details
| Gene Symbol: Ggnbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004273.1 | NP_001004273.1 | 12 | 2567 | ||
| XM_006247049.2 | XP_006247111.1 | 14 | 2778 | |||
| XM_006247050.2 | XP_006247112.1 | 14 | 2771 | |||
| XM_006247053.2 | XP_006247115.1 | 13 | 2650 | |||
| XM_006247054.2 | XP_006247116.1 | 13 | 2643 | |||
| GenBank mRNA | AY633742.1 | - | 12 | 2567 | ||

Ggnbp2
Genomic Map