siRNA Details
| Gene Symbol: Cops4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004275.1 | NP_001004275.1 | 2 | 176 | ||
| XM_006250668.2 | XP_006250730.1 | 2 | 222 | |||
| GenBank mRNA | BC079384.1 | - | 2 | 176 | 7111425 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cops4| Gene Symbol: Cops4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004275.1 | NP_001004275.1 | 2 | 176 | ||
| XM_006250668.2 | XP_006250730.1 | 2 | 222 | |||
| GenBank mRNA | BC079384.1 | - | 2 | 176 | 7111425 | |