siRNA Details
| Gene Symbol: Commd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004276.1 | NP_001004276.1 | 6 | 553 | ||
| XM_006254697.2 | XP_006254759.1 | 5 | 506 | |||
| GenBank mRNA | BC079373.1 | - | 6 | 553 | 7110105 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Commd10| Gene Symbol: Commd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004276.1 | NP_001004276.1 | 6 | 553 | ||
| XM_006254697.2 | XP_006254759.1 | 5 | 506 | |||
| GenBank mRNA | BC079373.1 | - | 6 | 553 | 7110105 | |