siRNA Details
| Gene Symbol: Pla2g15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004277.2 | NP_001004277.1 | 2 | 183 | ||
| GenBank mRNA | AY490816.1 | - | 2 | 155 | ||
| BC098894.1 | - | 2 | 183 | 7099635 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g15| Gene Symbol: Pla2g15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004277.2 | NP_001004277.1 | 2 | 183 | ||
| GenBank mRNA | AY490816.1 | - | 2 | 155 | ||
| BC098894.1 | - | 2 | 183 | 7099635 | ||