siRNA Details
| Gene Symbol: Tsga10ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004278.3 | NP_001004278.2 | 8 | 1773 | ||
| NM_001270736.1 | NP_001257665.1 | 9 | 1659 | |||
| NM_001270737.1 | NP_001257666.1 | 8 | 1647 | |||
| XM_006230847.2 | XP_006230909.1 | 9 | 1973 | |||
| XM_008760155.1 | XP_008758377.1 | 9 | 1983 | |||
| XM_008760156.1 | XP_008758378.1 | 8 | 1931 | |||
| XM_008760157.1 | XP_008758379.1 | 7 | 1762 | |||
| GenBank mRNA | AY690667.1 | - | 8 | 1789 | ||
| BC097254.1 | - | 8 | 1628 | 7453704 | ||

Tsga10ip
Genomic Map