siRNA Details
| Gene Symbol: Mmadhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004280.1 | NP_001004280.1 | 5 | 499 | ||
| XM_006234158.2 | XP_006234220.1 | 5 | 618 | |||
| GenBank mRNA | BC078953.1 | - | 5 | 499 | 7110154 | |
| FQ212704.1 | - | 5 | 494 | |||
| FQ213711.1 | - | 5 | 476 | |||
| FQ216239.1 | - | 5 | 494 | |||
| FQ219117.1 | - | 5 | 495 | |||
| FQ228704.1 | - | 5 | 495 | |||
| FQ229971.1 | - | 5 | 464 | |||

Mmadhc
Genomic Map