siRNA Details
| Gene Symbol: Eif3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004283.1 | NP_001004283.1 | 2 | 190 | ||
| XM_006241938.2 | XP_006242000.1 | 2 | 304 | |||
| GenBank mRNA | BC079005.1 | - | 2 | 190 | 7113045 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif3d| Gene Symbol: Eif3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004283.1 | NP_001004283.1 | 2 | 190 | ||
| XM_006241938.2 | XP_006242000.1 | 2 | 304 | |||
| GenBank mRNA | BC079005.1 | - | 2 | 190 | 7113045 | |