siRNA Details
| Gene Symbol: MIR646HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128288.1 | - | 2 | 4091 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR646HG| Gene Symbol: MIR646HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128288.1 | - | 2 | 4091 | ||