siRNA Details
| Gene Symbol: SLC22A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004326.4 | NP_001004326.4 | 5 | 894 | ||
| NR_033396.1 | 6 | 1063 | ||||
| XM_011545043.2 | XP_011543345.1 | 5 | 894 | |||
| XM_017017774.1 | XP_016873263.1 | 5 | 894 | |||
| XM_017017775.1 | XP_016873264.1 | 3 | 870 | |||
| XM_017017776.1 | XP_016873265.1 | 3 | 870 | |||
| GenBank mRNA | AK131327.1 | - | 5 | 896 | ||
| BC146850.1 | - | 5 | 886 | 9056778 | ||
| BM826695.1 | - | 2 | 226 | |||
| DN831899.1 | - | 2 | 158 | |||
| DY654969.1 | - | 3 | 333 | |||

SLC22A20
Genomic Map