siRNA Details
Gene Symbol: ZNF705A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004328.2 | NP_001004328.1 | 5 | 2941 |
Gene Symbol: ZNF705E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278713.1 | NP_001265642.1 | 5 | 2940 |
