siRNA Details
Gene Symbol: Tacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004415.2 | NP_001004415.1 | 5541 | |||
NM_001004418.2 | NP_001004418.1 | 426 | ||||
XM_008759884.1 | XP_008758106.1 | 6404 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004415.2 | NP_001004415.1 | 5541 | |||
NM_001004418.2 | NP_001004418.1 | 426 | ||||
XM_008759884.1 | XP_008758106.1 | 6404 |