siRNA Details
| Gene Symbol: MUC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040105.1 | NP_001035194.1 | 3 | 675 | ||
| NR_133665.1 | 3 | 675 | ||||
| GenBank mRNA | AJ606307.1 | - | 3 | 675 | ||
| AJ606308.1 | - | 3 | 675 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MUC17| Gene Symbol: MUC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040105.1 | NP_001035194.1 | 3 | 675 | ||
| NR_133665.1 | 3 | 675 | ||||
| GenBank mRNA | AJ606307.1 | - | 3 | 675 | ||
| AJ606308.1 | - | 3 | 675 | |||