siRNA Details
| Gene Symbol: Wapal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004436.4 | NP_001004436.2 | 3 | 1409 | ||
| NM_001301330.1 | NP_001288259.1 | 3 | 1409 | |||
| GenBank mRNA | AB167349.1 | - | 3 | 1215 | ||
| AK041191.1 | - | 3 | 1143 | |||
| AK045727.1 | - | 1 | 1727 | |||
| AK129105.1 | - | 3 | 1409 | |||
| BC038072.1 | - | 2 | 535 | |||
| BC139434.1 | - | 3 | 1164 | 8862456 | ||
| BC145625.1 | - | 3 | 1164 | 9054193 | ||

Wapal
Genomic Map