siRNA Details
| Gene Symbol: Pdia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004442.1 | NP_001004442.1 | 3, 4 | 246 | ||
| GenBank mRNA | BC082063.1 | - | 3, 4 | 246 | 7132589 | |
| X79328.1 | - | 2, 3 | 186 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdia6| Gene Symbol: Pdia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004442.1 | NP_001004442.1 | 3, 4 | 246 | ||
| GenBank mRNA | BC082063.1 | - | 3, 4 | 246 | 7132589 | |
| X79328.1 | - | 2, 3 | 186 | |||