siRNA Details
| Gene Symbol: Pdia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004442.1 | NP_001004442.1 | 6 | 538 | ||
| GenBank mRNA | BC082063.1 | - | 6 | 538 | 7132589 | |
| X79328.1 | - | 5 | 478 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdia6| Gene Symbol: Pdia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004442.1 | NP_001004442.1 | 6 | 538 | ||
| GenBank mRNA | BC082063.1 | - | 6 | 538 | 7132589 | |
| X79328.1 | - | 5 | 478 | |||