siRNA Details
| Gene Symbol: Rnf113a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004445.2 | NP_001004445.1 | 1 | 522 | ||
| GenBank mRNA | AY363109.1 | - | 1 | 603 | ||
| BC100134.1 | - | 1 | 522 | 7133304 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf113a2| Gene Symbol: Rnf113a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004445.2 | NP_001004445.1 | 1 | 522 | ||
| GenBank mRNA | AY363109.1 | - | 1 | 603 | ||
| BC100134.1 | - | 1 | 522 | 7133304 | ||