siRNA Details
| Gene Symbol: Smarca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004446.1 | NP_001004446.1 | 2, 3 | 384 | ||
| XM_006231227.2 | XP_006231289.1 | 2, 3 | 402 | |||
| XM_008760331.1 | XP_008758553.1 | 2, 3 | 662 | |||
| XM_008760332.1 | XP_008758554.1 | 3, 4 | 710 | |||
| XM_008760333.1 | XP_008758555.1 | 3, 4 | 450 | |||
| XM_008760334.1 | XP_008758556.1 | 2, 3 | 482 | |||
| XM_008760335.1 | XP_008758557.1 | 2, 3 | 418 | |||
| XM_008760336.1 | XP_008758558.1 | 2, 3 | 730 | |||
| GenBank mRNA | AY643746.1 | - | 2, 3 | 384 | ||

Smarca2
Genomic Map