siRNA Details
| Gene Symbol: Smarca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004446.1 | NP_001004446.1 | 3, 4 | 504 | ||
| GenBank mRNA | AY643746.1 | - | 3, 4 | 504 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smarca2| Gene Symbol: Smarca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004446.1 | NP_001004446.1 | 3, 4 | 504 | ||
| GenBank mRNA | AY643746.1 | - | 3, 4 | 504 | ||