siRNA Details
Gene Symbol: OR10J5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004469.1 | NP_001004469.1 | 1 | 818 | ||
GenBank mRNA | BC137025.1 | - | 1 | 863 | 9021020 | |
BC137026.1 | - | 1 | 863 | 9021021 |
