siRNA Details
| Gene Symbol: ST8SIA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004470.1 | NP_001004470.1 | 7 | 661 | ||
| XM_017016174.1 | XP_016871663.1 | 6 | 653 | |||
| GenBank mRNA | AB062101.1 | - | 7 | 661 | ||
| AJ621583.1 | - | 7 | 661 | |||
| BC137102.1 | - | 7 | 793 | 9021099 | ||
| BC137103.1 | - | 7 | 793 | 9021100 | ||
| BC144472.1 | - | 6 | 680 | |||

ST8SIA6
Genomic Map