siRNA Details
| Gene Symbol: ST8SIA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004470.1 | NP_001004470.1 | 6 | 536 | ||
| XM_017016174.1 | XP_016871663.1 | 5 | 528 | |||
| GenBank mRNA | AB062101.1 | - | 6 | 536 | ||
| AJ621583.1 | - | 6 | 536 | |||
| BC137102.1 | - | 6 | 668 | 9021099 | ||
| BC137103.1 | - | 6 | 668 | 9021100 | ||

ST8SIA6
Genomic Map