siRNA Details
| Gene Symbol: OR2T10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004693.1 | NP_001004693.1 | 1 | 643 | ||
| XM_017000273.1 | XP_016855762.1 | 2 | 894 | |||
| GenBank mRNA | BC136937.1 | - | 1 | 674 | 9020929 | |
| BC136940.1 | - | 1 | 674 | 9020932 | ||

OR2T10
Genomic Map