siRNA Details
| Gene Symbol: OR51F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004753.1 | NP_001004753.1 | 1 | 207 | ||
| GenBank mRNA | BC136877.1 | - | 1 | 272 | 9020868 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR51F2| Gene Symbol: OR51F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004753.1 | NP_001004753.1 | 1 | 207 | ||
| GenBank mRNA | BC136877.1 | - | 1 | 272 | 9020868 | |